ContactUsLinkComponent
Cap
9 mm Short-Cap, Screw-Vial Closures, Polypropylene, Preassembled, Restek
# de catalogue 10059-170
Fournisseur:  Restek
Cap
9 mm Short-Cap, Screw-Vial Closures, Polypropylene, Preassembled, Restek
# de catalogue 10059-170
Fournisseur:  Restek
Spécifications
  • Color:
    Red
  • Description:
    Short Screw Caps
  • Cat. no.:
    10059-170
  • Supplier No.:
    24503
  • Orifice:
    9 mm
Spécifications
A propos de cet article
9 mm short-cap closures are compatible with screw-thread or ribbed designs.
  • Economical
  • Sturdy polypropylene construction
These screw caps are for use with vials featuring short external threads where standard-length caps would be too long.
Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.
Customers Who Bought This Also Bought