ContactUsLinkComponent
Decapper Easy Grip Manual Crimper Size:20 mm
Easy Grip Manual Crimpers and Decappers, Restek
# de catalogue 10059-100
Fournisseur:  Restek
Decapper Easy Grip Manual Crimper Size:20 mm
Easy Grip Manual Crimpers and Decappers, Restek
# de catalogue 10059-100
Fournisseur:  Restek
Spécifications
  • Description:
    Decapper
  • Cat. no.:
    10059-100
  • Seal Size:
    20 mm
  • Supplier No.:
    23399
Spécifications
A propos de cet article
Restek's Easy Grip crimpers and decappers feature tough, light, plastic bodies with curved handles that improve hand comfort during use.
The bottom-pull handle allows for a steadier hold than top-push handles. An easily viewed adjustment knob is located at the head of the product. The clearly shown + and – symbols, along with directional arrows, simplify adjusting the intensity of the crimp/decap as needed. The knob is also designed to indicate when the crimp setting has been reached, when it fully contacts the body of the product.
Customers Who Bought This Also Bought