HyperSep Glass Block Manifolds, Thermo Scientific
# de catalogue 10047-016
Fournisseur: NATIONAL SCIENTIFIC MX
Spécifications
- Description:24-Port Vacuum Manifold
- Cat. no.:10047-016
- Supplier No.:60104-233
Spécifications
A propos de cet article
Provides multiple processing of SPE or SLE cartridges to improve throughput and reproducibility.
Delivery: Includes glass block manifold lid, cover gasket, vacuum gauge, valve assembly, tips (16 or 24), adjustable collection rack, bulkhead luer fittings, plugs (16 or 24) and manifold safety tray.
Customers Who Bought This Also Bought