Acclaim™ C30 LC Columns, Thermo Scientific
Catalogue # 10044-772
Fournisseur: Thermo Scientific
Spécifications
- Column Format:Analytical
- Ø int.:4.60 mm
- Particle Size:5 µm
- Pore size:200 Å
- Length:250 mm
- pH range:2 to 8
- Surface area:200 m²/g
- Carbon load:13%
- Description:10044-772
- Cat. no.:10044-772
- Supplier No.:075718
Spécifications
À propos de cet article
For separations of hydrophobic, structurally related isomers, Thermo Scientific™ Acclaim C30 reversed-phase columns provide superior results.
The covalent modification of C30 alkyl silane on ultrapure, spherical, porous silica gel gives these columns high shape selectivity. They are fully compatible with various aqueous buffers, useful for a broad application range of analytes, and offer more flexibility in method development.
Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (Part No. 10046-750).
Frequently Bought Together