ContactUsLinkComponent
WEBSEAL MAT 96 Round FLAT 7MM DRK BLUE T/S Dark Blue T/S, 5/Pk
WebSeal™ Standard Mats and Sealing Tapes, Non sterile
# de catalogue 10036-242
Fournisseur:  Thermo Scientific
WebSeal™ Standard Mats and Sealing Tapes, Non sterile
# de catalogue 10036-242
Fournisseur:  Thermo Scientific
Spécifications
  • Color:
    Blue
  • Description:
    WebSeal Mats, 96-Well, Flat Base, Round Well, 7 mm
  • Material:
    Silicone
  • Supplier No.:
    60180-M111
Spécifications
A propos de cet article
These sealing mats are placed directly on the microplate by hand or with an applicator.
Manufactured from pure silicone with wider or small penetration area for ease of use. Mats are dry heat autoclavable, provide excellent chemical compatibility, and resist coring and tearing. Pre-slit mats ensure easy penetration and reduce vacuum problems. Non sterile.
Customers Who Bought This Also Bought