ContactUsLinkComponent
CERT well Plate 96 7MM Round U BASE 1.0ML PP U Base, 1.0Ml, Pp, 50/Cs
Certified WebSeal™ Plates, Plastic, Non-Coated, Nonsterile, Thermo Scientific
# de catalogue 10036-216
Fournisseur:  Thermo Scientific
Certified WebSeal™ Plates, Plastic, Non-Coated, Nonsterile, Thermo Scientific
# de catalogue 10036-216
Fournisseur:  Thermo Scientific
Spécifications
  • Volume:
    1000 µl
  • Description:
    96-Well, Deep Well Microplates, Round Wells
  • Height:
    41.6 mm
  • Well Shape:
    U-Shaped
  • Supplier No.:
    60180-P201
Spécifications
A propos de cet article
Manufactured from ultra-low bleed, high-purity basis resin
Fully lot tested by GC-MS and LC/MS for organic extractables.
Customers Who Bought This Also Bought