Certified WebSeal™ Plates, Plastic, Non-Coated, Nonsterile, Thermo Scientific
# de catalogue 10036-210
Fournisseur: Thermo Scientific
Spécifications
- Volume:270 µl
- Description:96-Well, Deep Well Microplates, Round Wells
- Height:14.7 mm
- Well Shape:U-Shaped
- Supplier No.:60180-P216
Spécifications
A propos de cet article
Manufactured from ultra-low bleed, high-purity basis resin
Fully lot tested by GC-MS and LC/MS for organic extractables.
Customers Who Bought This Also Bought