Precision Zip-Clean Pouches™, PPC Flexible Packaging
Catalog # 10033-172
Supplier: Fisher Container
Specifications
- Dimensions:45.7×61 cm (18×24")
- Cat. no.:10033-172
- Supplier No.:91090P
Specifications
About this item
Tough, ultraclean, high clarity film that is ideal for all multi-use, reclosable cleanroom and laboratory applications.
- Easy open and easy zip closure
- Cleanroom certified
- Barrier properties
Sealed in a double-lined carton.
Customers Who Bought This Also Bought