Precision Zip-Clean Pouches™, PPC Flexible Packaging
Catalog # 10033-170
Supplier: Fisher Container
Specifications
- Dimensions:30.5×38.1 cm (12×15")
- Cat. no.:10033-170
- Supplier No.:91089P
Specifications
About this item
Tough, ultraclean, high clarity film that is ideal for all multi-use, reclosable cleanroom and laboratory applications.
- Easy open and easy zip closure
- Cleanroom certified
- Barrier properties
Sealed in a double-lined carton.
Customers Who Bought This Also Bought