Precision Zip-Clean Pouches™, PPC Flexible Packaging
# de catalogue 10033-168
Fournisseur: Fisher Container
Spécifications
- Dimensions:25.4×30.5 cm (10×12")
- Cat. no.:10033-168
- Supplier No.:91088P
Spécifications
A propos de cet article
Tough, ultraclean, high clarity film that is ideal for all multi-use, reclosable cleanroom and laboratory applications.
- Easy open and easy zip closure
- Cleanroom certified
- Barrier properties
Sealed in a double-lined carton.
Customers Who Bought This Also Bought