Certified Reference Materials, Sulphide, Rocklabs
# de catalogue CA10032-606
Fournisseur: ROCKLABS (W/T) CA
Numéro CAS:
Spécifications
- Packaging:Jar
- Description:SJ Sulphide (3.0% S) 2.00 - 3.00 ppm Au
- Size:2.5 kg
- Cat. no.:CA10032-606
- Supplier No.:301860
Spécifications
A propos de cet article
Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag
Recommended Gold (ppm) concentrations. Packaged in jars or satchets.
Customers Who Bought This Also Bought