Accessories for VWR® Forced Air Microbiological Incubators
# de catalogue 10031-770
Fournisseur: VWR International
Spécifications
- Description:Extra Perforated Shelf with Supports for VWR® Large Forced Air Incubator (6.3 cu. ft.), 17.28×19.88", 7 lbs.
- Cat. no.:10031-770