Tite-Rite™ Containers, Globe Scientific
# de catalogue 10030-306
Fournisseur: Globe Scientific
Spécifications
- Cap Size:53 mm
- Capacity:120 ml (4 oz.)
- Material:PP
- Supplier No.:6527
Spécifications
A propos de cet article
Tite-Rite™ containers provide the ultimate convenience and reliability in leak-tight storage and liquid transport
Manufactured from polypropylene, these conatiners are autoclavable at 121°C (250°F) for 20 minutes (without caps). Containers are 95kPa compliant. Caps are manufactured from polyethylene.
Manufactured from polypropylene, these conatiners are autoclavable at 121°C (250°F) for 20 minutes (without caps). Containers are 95kPa compliant. Caps are manufactured from polyethylene.
The specially designed caps contain an inner liner designed to compress against the uppermost walls of the container, ensuring an air-tight seal. The containers are ideal for use in hospital pneumatic tube systems.
Customers Who Bought This Also Bought