Water, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
# de catalogue BDH23595.100
Fournisseur: VWR International
Numéro CAS: 7732-18-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber
- Size:1 L
- Cat. no.:BDH23595.100
- Supplier No.:VWRCBDH23595.1
Spécifications
Résultats des tests de spécification
- Filtered at 0.2 µm:Passes Test
- Coloration:Max. 10 APHA
- Evaporation Residue:Max. 1 ppm
- Spec. Conductivity (25°C) (at filling):Max. 1 µS/cm
- Total Organic Carbon (TOC):Max. 10 ppb
- Absorbance (200 nm):Max. 0.01
- Absorbance (210 nm):Max. 0.01
- Absorbance (220 nm):Max. 0.01
- Absorbance (240 nm):Max. 0.01
- Absorbance (254 nm):Max. 0.005
- Absorbance (280 nm):Max. 0.005
- Absorbance (300 nm):Max. 0.005
- Fluorescence (as quinine) (254 nm):Max. 1 ppb
- Fluorescence (as quinine) (365 nm):Max. 0.5 ppb
- Gradient Grade (210 nm):Max. 5 mAU
- Gradient Grade (254 nm):Max. 1 mAU
- VWR Chemicals BDH®, Water, HiPerSolv CHROMANORM® for HPLC:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydrogen oxide
:Eau
:Storage Temperature: Ambient
Point d'ébullition:100 °C (1013 hPa)
Point de Fusion:0 °C
Densité:1 g/cm³ (4 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Point d'ébullition:
:Pt ébul.: 100 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: 0 °C
:Point eclaire:
:Flash Pt:
:M.W. 18,02 g/mol
Index Merck:13,10098
EINECS:231-791-2
:0
:100
Température d'entreposage:Température ambiante
:12,10175
Customers Who Bought This Also Bought