ContactUsLinkComponent
IATA-B Refrigerated Shipper, 3/Cs
Shipper, Category B Refrigerated, Medium 95 kPa Canister, Therapak®
# de catalogue 10029-298
Fournisseur:  Therapak, LLC
IATA-B Refrigerated Shipper, 3/Cs
Shipper, Category B Refrigerated, Medium 95 kPa Canister, Therapak®
# de catalogue 10029-298
Fournisseur:  Therapak, LLC
Spécifications
  • Description:
    Cat B refrigerated, Medium 95kpa canister
  • Cat. no.:
    10029-298
  • Supplier No.:
    37815
Spécifications
A propos de cet article
Refrigerated Shipping System includes transport box (OD: 11x9x11"), small cooler (ID: 8x6x8"), medium 95kPa canister, Aqui-Pak™ segmented absorbent pouch, 8 oz
refrigerant packs, cushioning material, list of contents card, and required labels for IATA PI650 marking and compliance. Holds up to six tubes for overnight shipment.
Customers Who Bought This Also Bought