Screw Caps for Sample Tubes, with O-Ring, Globe Scientific
# de catalogue 10028-126
Fournisseur: Globe Scientific
Spécifications
- Color:Gray
- Cat. no.:10028-126
- Supplier No.:6137A
Spécifications
A propos de cet article
All caps include an O-Ring to ensure a leak-tight seal at most temperatures.
Customers Who Bought This Also Bought