ContactUsLinkComponent
Flip & Grippipette Holder
Flip & Grip™ Reversible Pipettor Holder, Heathrow Scientific®
# de catalogue 89511-784
Fournisseur:  VWR International
Flip & Grippipette Holder
Flip & Grip™ Reversible Pipettor Holder, Heathrow Scientific®
# de catalogue 89511-784
Fournisseur:  VWR International
Spécifications
  • Color:
    Lime
  • Description:
    Reversible Pipettor Holder
  • Cat. no.:
    89511-784
  • Supplier No.:
    HS120056
Spécifications
A propos de cet article
This unique pipettor holder allows users the option of use on the benchtop, or hanging under a shelf to maximize available working space
The benchtop style measures 332 Hx193 W mm, and the under shelf style measures 187 H x 193 W mm.
Each Flip & Grip™ holder will accommodate up to 12 single channel pipettors, or a combination of 3 single and 3 multichannel pipettors. The unit holds all the most common pipettor brands. Rubber grips prevent scratching. The holder rotates for easy selection and storage. Available in three colors.
Ordering information: Mounting hardware included.
Customers Who Bought This Also Bought