Dichloromethane ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
# de catalogue BDH23373.400
Fournisseur: VWR International
Product available on GSA Advantage®
Numéro CAS: 75-09-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber
- Size:4 L
- Supplier No.:VWRCBDH23373.4
Spécifications
Résultats des tests de spécification
- Assay (GC):Min. 99.8 %
- Water:Max. 0.05 %
- Non-volatile residue:Max. 0.001 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Transmittance (240 nm):Min. 50 %
- Transmittance (245 nm):Min. 80 %
- Transmittance (260 nm):Min. 98 %
- Conforms to BDH 15245:Passes test
- VWR Chemicals BDH®, Dichloromethane ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC:
Résultats des tests de spécification
A propos de cet article
Synonymes: Methylene chloride
:Storage Temperature: Ambient
Point d'ébullition:39,8 °C (1013 hPa)
Point de Fusion:–95 °C
Densité:1,322 g/cm³ (20 °C)
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Filtré à 0,2 µm. Conditionné sous azote.
:stabilisé avec 2-méthyl-2-butène 20 ppm
:Point d'ébullition:
:Pt ébul.: 39,8 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: -95 °C
:Point eclaire:
:Flash Pt:
:M.W. 84,93 g/mol
Index Merck:12,06140
:Dichlorométhane
EINECS:200-838-9
:-95
:39,8
ONU:1593
:6.1
:III
:GHS08
:GHS07
Température d'entreposage:Température ambiante
:2-Methyl-2-buten
:20 ppm
:Filter 0,2 um_Nitrogen_filled
Customers Who Bought This Also Bought