ContactUsLinkComponent
Rack For 50Ml Centrifuge Tube Orange Cs 50 Pk 5 Sterile
VWR® Standard Line Sterile Racks for 15 and 50 ml Conical-Bottom Tubes, PP
# de catalogue 10026-312
Fournisseur:  VWR International
VWR® Standard Line Sterile Racks for 15 and 50 ml Conical-Bottom Tubes, PP
# de catalogue 10026-312
Fournisseur:  VWR International
Spécifications
  • Color:
    Orange
  • Description:
    Racks for 50 ml Tubes
  • Cat. no.:
    10026-312
Spécifications
A propos de cet article
Polypropylene racks are available in seven colors, and are also offered in an assorted color pack with one of each color: blue, green, orange, red, and yellow.
  • Available in a variety of colors
  • Suitable for plug or flat cap tubes
  • Polypropylene construction
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Racks accommodate plug or flat cap tubes.
Certifications: TSE/BSE-free Certification.
Ordering information: All racks are sterilized by irradiation SAL10-6, and supplied RNase-free, DNase-free, and non-pyrogenic.
Customers Who Bought This Also Bought