Order Entry
ContactUsLinkComponent
Blocker* Casein in PBS 1L
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
# de catalogue CAPI37528
Fournisseur:  Thermo Scientific
Numéro CAS:  
Blocker* Casein in PBS 1L
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
# de catalogue CAPI37528
Fournisseur:  Thermo Scientific
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Casein in PBS
  • Size:
    1 L
  • Cat. no.:
    CAPI37528
  • Application:
    ELISA, Western Blot, Dot Blot, Immunohistochemistry, Hybridization
  • Supplier No.:
    37528

Spécifications

A propos de cet article

Thermo Scientific Blocker Casein in PBS or TBS is a ready-to-use, solution of purified casein protein for blocking steps in Western blot, ELISA, IHC and nucleic acid detection methods.

  • Easy to use
  • Safe, stable and thimerosal-free formulations
  • Purified casein provides fewer chances of cross reaction
  • Flexible and available in PBS and TBS formulations

This blocking buffer contains casein protein that is purified from milk by the Hammarsten method. The phosphate- and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody-based detection methods. Blocker Casein Buffers are 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought