Single Probe Thermometers with Alarm, Time Date Stamp, Thermco
Catalog # 10025-452
Supplier: Thermco
Specifications
- Description:Thermometer, Incubators, certified at 37 °C
- Cat. no.:10025-452
- Probe:One 5 ml bottle, Bio-Safe glycol/water
- Range:–50...+70 °C (...58...+158 °F)
- Supplier No.:ACC821INCV
Specifications
About this item
These large digital thermometers simultaneously show the current, minimum, and maximum temperatures and update continuously.
- Triple display
- Visual LED and audible beeping alarms
- Bio-safe glycol/water probe medium
- Improved resolution to 0.1º
- Standard size vaccine bottles
- Min.-max. temp with time and date stamp
The probe is housed in an unbreakable plastic bottle filled with a buffering media that maintains the thermometer reading when the instrument door is opened. The thermometer and the probe bottle have dual magnets to attach to the instrument. Thermometers also feature a flip-open stand, wall mounting key hole, and an AAA alkaline battery.
An audible and visual alarm will trigger when the temperatures goes above or below the set temperatures. The minimum/maximum feature monitors critical temperature for any period of time.
Certifications: Each thermometer is certified and traceable to NIST. Each thermometer is serialized.
Customers Who Bought This Also Bought