Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
# de catalogue BDH20864.400
Fournisseur: VWR International
Product available on GSA Advantage®
Numéro CAS: 67-56-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Supplier No.:VWRCBDH20864.4
Spécifications
Résultats des tests de spécification
- Assay (on anhydrous substance):Min. 99.8 %
- Acidity:Max. 0.0003 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Evaporation residue:Max. 5 ppm
- Water:Max. 0.03 %
- Transmittance (220 nm):Min. 50 %
- Transmittance (235 nm):Min. 80 %
- Transmittance (260 nm):Min. 98 %
- VWR Chemicals BDH®, Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydroxy methane,Carbinol,Wood alcohol
Formule:CH₃OH
Point d'ébullition:64,6 °C (1013 hPa)
Point de Fusion:–98 °C
Densité:0,7918 g/cm³ (20 °C)
Point d'éclair:11 °C (closed cup)
Numéro MDL:MFCD00004595
Numéro CAS:67-56-1
:Storage Temperature: Ambient
ADR:3,II
:Length
:Width
:Height
:Diameter
:Thickness
:99,8
:Filtré à 0,2 µm. Conditionné sous azote.
:Point d'ébullition:
:Pt ébul.: 64,6 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: -98 °C
:Point eclaire:
:Pt Eclair: 11 °C (coupe fermée)
:M.W. 32,04 g/mol
Index Merck:13,05984
EINECS:200-659-6
:-98 °C
:64,6
ONU:1230
:01-2119433307-44
:3
:II
:GHS02
:GHS06
:GHS08
Température d'entreposage:Température ambiante
:Filter 0,2 um_Nitrogen_filled
Customers Who Bought This Also Bought