ContactUsLinkComponent
VWR® Heat Resistant Film for Thermal Cycling
Catalogue # 89496-565
Fournisseur:  VWR International
undefined
VWR® Heat Resistant Film for Thermal Cycling
Catalogue # 89496-565
Fournisseur:  VWR International
Spécifications
  • Sterility:
    Non sterile
  • Thickness:
    50 µm
  • Cat. no.:
    89496-565
Spécifications
À propos de cet article
VWR® Heat Resistant Films are improved films for classic thermal cycling applications.
  • Improved adhesion
  • Reduced evaporation
  • Longer end tabs for easier handling
  • Heat-resistant
They are made of 50 Uu Polyester with a 41 Um layer of acrylic adhesive
VWR® Heat Resistant Films are recommended for temperatures from –40 to 120°C. Films are certified DNase-, RNase-, and nucleic acid-free.
The films offer better adhesion and reduced evaporation. They feature longer end tabs for easier handling.
Frequently Bought Together