Specifications
- Volume:100 µL
- Tip Color:Transparent
- Packed:96/tray
- Cat. no.:CAPI87784
- Tip Style:C18
- For Use With:Mass spectrometer, microcentrifuge
- Supplier No.:87784
Specifications
About this item
Pierce™ C18 Tips are ready-to-use pipette-tip columns of C18 resin that enable fast and efficient capture, concentration, desalting and elution of peptides for MALDI mass spectrometry and other methods.
- Monolithic C18 fast-flow tips have low resistance and improved flow characteristics compared to other commercially available tips
- Eight packs and 96-tip trays are automation compatible
Improve protein analysis results with Pierce™ C18 Tips by removing urea, salts and other contaminants before MS analysis.
These C18 tips offer excellent flow properties with a high-efficiency monolithic C18 sorbent for fast wetting, loading, washing and eluting. The five-step procedure is simple and requires less than 5 minutes to process protein digests, strong cation exchange fractions and other protein and peptide samples for mass spectrometric (MS) analyses.
The tips are ideal for preparing samples for matrix-assisted laser desorption ionization (MALDI) or nanoelectrospray ionization techniques. The Pierce™ C18 Tips are available in convenient 10 and 100 µl tips with binding capacities of 8 and 80 µg, respectively.