Accessories for PIPETBOY acu 2 Pipette Aid, Integra Biosciences
Catalog # 89498-824
Supplier: INTEGRA Biosciences
Specifications
- Description:Filter, Sterile (0.45μm pore size)
- Cat. no.:89498-824
- Supplier No.:153015