Polystyrene and Polypropylene Test Tubes, 13×100 mm, Globe Scientific
# de catalogue 89497-824
Fournisseur: Globe Scientific
Spécifications
- Diameter ×L:13 x 100 mm
- Volume:8 ml
- Description:Test Tube
- Material:PP
- Supplier No.:110445
Spécifications
A propos de cet article
These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.
Customers Who Bought This Also Bought