Polystyrene and Polypropylene Test Tubes, 13×100 mm, Globe Scientific
Catalog # 89497-808
Supplier: Globe Scientific
Specifications
- Diameter ×L:13×100 mm
- Volume:8 ml
- Description:Test Tube
- Material:Polystyrene
- Supplier No.:110415
Specifications
About this item
These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.
Frequently Bought Together