Polystyrene and Polypropylene Test Tubes, 17×100 mm, Globe Scientific
Catalog # 89497-788
Supplier: Globe Scientific
Specifications
- Ø×L:17 x 100 mm
- Volume:Test tube
- Description:Test Tube
- Material:Polystyrene
- Cat. no.:89497-788
- Supplier No.:110182
Specifications
About this item
These tubes are molded either from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis and solvents.
Caps are made of polyethylene and are not autoclavable.
Customers Who Bought This Also Bought