Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
Catalogue # 52458-147
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Spécifications
- Description:1413 µS/cm Conductivity Standard, 692 ppm as NaCl TDS Standard
- Size:5×60 ml
- Cat. no.:52458-147
- Supplier No.:011007
Spécifications
À propos de cet article
Perform virtually any electrochemistry analysis with Orion™ Conductivity Standards and Solutions
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
Conductivity standard temperature tables are programmed into Orion™ meter software for use with the automatic calibration feature.
Choose from a complete selection of conductivity accessories including conductivity standards and TDS (total dissolved solids) standards. All conductivity standards are traceable to NIST standard reference materials.
Frequently Bought Together