Slippable Petri Dishes, 100 mm
# de catalogue 470210-568
Fournisseur: VWR International
Spécifications
- Dimensions:100×15 mm
- Sterility:Sterile
- Description:Petri dish, slippable
- Cat. no.:470210-568
Spécifications
A propos de cet article
These slippable polystyrene Petri dishes provide optimal flatness.
- Unsurpassed in optical calrity - Ideal for micro photography
- Engineered for optimum flatness to provide uniform agar thickness
- Manufactured from pure virgin polystyrene
Offered in a convenient pack of six for small batches.
Packaging: Pack of six.
Customers Who Bought This Also Bought