VWR® Filter Holder 47 mm, 40/35 Joint Connection
# de catalogue 89428-970
Fournisseur: VWR International
Spécifications
- Description:Filter Holder with Glass Support
- Cat. no.:89428-970
Spécifications
A propos de cet article
This unit is supplied with a 300 ml graduated funnel, an anodized aluminum clamp, and a 1000 ml flask.
- Borosilicate glass fabrication
- Two types of holders available
Glass components of this unit are manufactured from Borosilicate 3.3 Glass.
Fritted glass support base or a 100 mesh stainless steel support screen, PTFE-support screen gasket, and glass support base.
Customers Who Bought This Also Bought