Accessories for VWR® Filter Holders 47 mm
Catalog # 89428-954
Supplier: VWR International
Specifications
- Description:VWR® Glass Funnel, 300 ml
- Cat. no.:89428-954
Specifications
About this item
Customers Who Bought This Also Bought
Accessories for Single-Sample Osmometer, Model 3250, Advanced Instruments
Supplier: Advanced Instruments
$554.99
Masterflex® L/S® Easy-Load® II Pump Heads for High-Performance Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
$979.60
VWR® Graduated Cylinders, Calibrated To Contain, Class B
Supplier: VWR International
$79.45 to $714.70
Dimethyl sulfoxide, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$1,638.10 to $5,103.86
Potassium chloride 99.0-100.5% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$219.92 to $686.98
Falcon™ Cell Strainers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$163.12 to $178.73