Accessories for VWR® Filter Holders, 25 mm
# de catalogue 89428-938
Fournisseur: VWR International
Spécifications
- Description:VWR® Glass Funnel, 15 ml
- Cat. no.:89428-938
Spécifications
A propos de cet article
Customers Who Bought This Also Bought
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Fournisseur: Saint Gobain Performance Plastics
$1,747.88
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
$362.33
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Fournisseur: VWR International
$541.80
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
$881.47
Phosphate Standard, 1 ml = 0.05 mg P, 50 ppm P (153 ppm PO₄³⁻), Ricca Chemical Company
Fournisseur: Ricca Chemical
$159.69
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrite (328.5 ppm NO₂⁻), Ricca Chemical Company
Fournisseur: Ricca Chemical
$145.60
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
$464.49