Automation Tips for Eppendorf® epMotion™ Automated Pipetting System, Thermo Scientific
Catalog # 89428-730
Supplier: Thermo Scientific
Specifications
- Volume:300 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Packed:96 Tips/Rack, 10 Racks/Pack, 4 Packs/Case
- Cat. no.:89428-730
- Tip Style:Standard
- Supplier No.:966-262
Specifications
About this item
Thermo Scientific provides a solution for your Eppendorf epMotion robotic pipette tips.
These automation pipette tips have been designed, validated, and tested on an actual epMotion workstation to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our epMotion workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Frequently Bought Together