Vacuum Storage Tube with Hi-Vac Top Valve and Outer Standard Taper Joint Sidearm, Ace Glass Incorporated
# de catalogue 89427-992
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Capacity:25 ml
- ST Joint No.:14/20
- Description:Vacuum Storage Tube with Hi-Vac Top Valve and Outer Standard Taper Joint Sidearm
- Cat. no.:89427-992
- Supplier No.:7821-01
Spécifications
A propos de cet article
This borosilicate glass vacuum storage tube features an 0-4mm Hi-Vac valve top with a standard taper jointed sidearm
The valved top enables precise flow control and the standard taper, outer joint facilitates connections to Schlenk lines.
Customers Who Bought This Also Bought