ContactUsLinkComponent
TUBESTORAGE25MLVLV14/20
Vacuum Storage Tube with Hi-Vac Top Valve and Outer Standard Taper Joint Sidearm, Ace Glass Incorporated
# de catalogue 89427-992
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
TUBESTORAGE25MLVLV14/20
Vacuum Storage Tube with Hi-Vac Top Valve and Outer Standard Taper Joint Sidearm, Ace Glass Incorporated
# de catalogue 89427-992
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Capacity:
    25 ml
  • ST Joint No.:
    14/20
  • Description:
    Vacuum Storage Tube with Hi-Vac Top Valve and Outer Standard Taper Joint Sidearm
  • Cat. no.:
    89427-992
  • Supplier No.:
    7821-01
Spécifications
A propos de cet article
This borosilicate glass vacuum storage tube features an 0-4mm Hi-Vac valve top with a standard taper jointed sidearm
The valved top enables precise flow control and the standard taper, outer joint facilitates connections to Schlenk lines.
Customers Who Bought This Also Bought