ContactUsLinkComponent
TUBESTORAGE210MLVALVECAJON
Vacuum Storage Tube, with Hi-Vac TopValve and ⅜" Cajon Sidearm, Ace Glass Incorporated
# de catalogue 89427-982
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
TUBESTORAGE210MLVALVECAJON
Vacuum Storage Tube, with Hi-Vac TopValve and ⅜" Cajon Sidearm, Ace Glass Incorporated
# de catalogue 89427-982
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Capacity:
    210 ml
  • Description:
    Vacuum Storage Tube, with Hi-Vac Top Valve and 3/8" Cajon Sidearm
  • Cat. no.:
    89427-982
  • Supplier No.:
    7813-15
Spécifications
A propos de cet article
This borosilicate storage tube is supplied with an 0-4mm Hi-Vac valve top with a 9mm side port designed to accept ⅜" Cajon fittings
The valved top enables precise flow control.
Customers Who Bought This Also Bought