Glass or PTFE Baffles, Ace Glass Incorporated
# de catalogue 89427-764
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Ø:6.4 mm
- Length:327 mm
- Description:PTFE Baffle
- Cat. no.:89427-764
- Ace-thred no.:#7
- Supplier No.:12177-145
Spécifications
A propos de cet article
Use Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint
Baffles can be adjusted vertically or rotated to increase agitation.
Customers Who Bought This Also Bought