ContactUsLinkComponent
Human Transferrin Cf488A
CF™ Dye Transferrin Conjugates, Biotium
# de catalogue 89427-096
Fournisseur:  Biotium
Numéro CAS:  
CF™ Dye Transferrin Conjugates, Biotium
# de catalogue 89427-096
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Human Transferrin, CF™488A Conjugate
  • Cat. no.:
    89427-096
  • Supplier No.:
    81
Spécifications
A propos de cet article
Transferrin is a glycoprotein with homologous N-terminal and C-terminal iron-binding domains that binds up to two Fe(III)atoms for delivery to vertebrate cells through receptor-mediated endocytosis
CF dye technology is covered by pending U.S. and international patents.
After binding to its receptor on the cell surface, transferrin is rapidly internalized by invagination of clathrin-coated pits with formation of endocytic vesicles, the acidic environment favors dissociation of iron from the transferrin-receptor complex. Following the release of iron, the apotransferrin is recycled to the plasma membrane where it is released from its receptor to scavenge more iron. The human transferrin is labeled with our bright and photostable CF dyes that aids the investigation of endocytosis by microscopy.
Customers Who Bought This Also Bought