Decontamination bags, activated charcoal, Biotium
Catalog # 89427-060
Supplier: Biotium
CAS Number:
Specifications
- Description:Activated Charcoal Decontamination Bags
- Size:25 bags
- Cat. no.:89427-060
- Supplier No.:22007
Specifications
About this item
Activated charcoal retains ethidium bromide, GelRed™ and GelGreen™ efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.
- Remove mutagenic dyes (ie, ethidium bromide) from gel staining solutions
- Use to clean up any dyes from a solution to allow drain disposal
- No-mess, convenient bag
- Reusable bag capacity of at least 10 mg EtBr
Frequently Bought Together