Round Bottom Flask, Double Neck, Glassco
Catalog # 89426-520
Supplier: Glassco
Specifications
- ST Joint No.:24/40
- Capacity:100 ml
- Description:Round Bottom Flask, Double Neck
- Cat. no.:89426-520
- Supplier No.:059.470.05A
Specifications
About this item
Round bottom flask with same sized joints
Heavy wall design. Manufactured from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform wall thickness.
Customers Who Bought This Also Bought