Round Bottom Flask, Single Neck, Glassco
# de catalogue 89426-472
Fournisseur: Glassco
Spécifications
- Capacity:50 ml
- ST Joint No.:19/22
- Description:Round Bottom Flask, Single Neck
- Supplier No.:057.470.09
Spécifications
A propos de cet article
Heavy wall design with single neck
Made from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform thickness.
Customers Who Bought This Also Bought