ContactUsLinkComponent
Flask Round Bottom 1N 24/40 1000Ml
Round Bottom Flask, Single Neck, Glassco
# de catalogue 89426-466
Fournisseur:  Glassco
Flask Round Bottom 1N 24/40 1000Ml
Round Bottom Flask, Single Neck, Glassco
# de catalogue 89426-466
Fournisseur:  Glassco
Spécifications
  • Capacity:
    1000 ml
  • ST Joint No.:
    24/40
  • Description:
    Round Bottom Flask, Single Neck
  • Supplier No.:
    057.202.61
Spécifications
A propos de cet article
Heavy wall design with single neck
Made from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform thickness.
Customers Who Bought This Also Bought