ContactUsLinkComponent
Adapter Vac Tfe Stpk 24/40
Vacuum Adapter, with PTFE Stopcock, Glassco
Catalog # 89426-408
Supplier:  Glassco
Adapter Vac Tfe Stpk 24/40
Vacuum Adapter, with PTFE Stopcock, Glassco
Catalog # 89426-408
Supplier:  Glassco
Specifications
  • Description:
    Vacuum Adapter, with PTFE Stopcock
  • Cat. no.:
    89426-408
  • ST Joint Size:
    24/40
  • Supplier No.:
    034.470.03J
Specifications
About this item
Vacuum adapters are designed to regulate the flow of gases or liquids into reaction systems.
  • PTFE stopcock
  • Hose connection
Stopcock has a bore of 2mm. Hose connection has an O.D. of 8mm at the middle serration with standard inner taper.
Customers Who Bought This Also Bought