Order Entry
ContactUsLinkComponent
VARIABLE TIMER/CONTROLLER
Heating Mantle Controller
# de catalogue 470202-750
Fournisseur:  Glas-Col
VARIABLE TIMER/CONTROLLER
Heating Mantle Controller
# de catalogue 470202-750
Fournisseur:  Glas-Col

Spécifications

  • Description:
    Heating Mantle Controller
  • Cat. no.:
    470202-750
  • Supplier No.:
    104A PL312

Spécifications

A propos de cet article

Regulates Heating Mantles, Tapes and Cords, Small Furnaces, Ovens and Hot Plates.

  • Analog controller
  • Provides 5.5% to 100% full power

Anodized aluminum case with fuse, pilot light, and 6' grounded line cord. Size: 4¼" L x 2½" W x 2" H. For 120V, 50/60Hz, 1500W. CSA listed.

Customers Who Bought This Also Bought