Order Entry
ContactUsLinkComponent
METRIC ADHESIVE TAPE18M RL (60 FT)PVC
Metric Adhesive Tape
# de catalogue 470201-356
Fournisseur:  xpedx
METRIC ADHESIVE TAPE18M RL (60 FT)PVC
Metric Adhesive Tape
# de catalogue 470201-356
Fournisseur:  xpedx

Spécifications

  • Cat. no.:
    470201-356
  • Supplier No.:
    5382711

Spécifications

A propos de cet article

Versatile adhesive tape with many classroom uses.

  • Graduated in mm and numbered in cm; repeats every 100 cm
Packaging: Supplied as 18 m roll.
Customers Who Bought This Also Bought