Glass Depression Slides
Catalog # 470200-930
Supplier: United Scientific Supplies
Specifications
- Description:Double cavity slides
- L×W:3×1"
- Thickness:1.3 mm
- Cat. no.:470200-930
- Supplier No.:CS3X12
Specifications
About this item
These depression slides are ideal for viewing living specimens suspended in liquid.
- Made with quality sheet glass with ground edges
- Sold in boxes of 12 slides
- Available with single or double concavities
These precleaned slides have a round, shallow depression in the center of the slide, making it easy to view materials in liquid.
Cavities measure approximately 16mm in diameter, and 0.8mm deep.
Customers Who Bought This Also Bought