Distilled Water
# de catalogue 470200-788
Fournisseur: MAYER HARDWARE SE
Numéro CAS:
Spécifications
- Description:Distilled Water
- Size:1 Gal
- Cat. no.:470200-788
- Grade:Laboratory
- Supplier No.:887005
Spécifications
A propos de cet article
CAS Number: 7732-18-5
Formula: H2O
Density: 1 g/mL
Boiling and Freezing Point: 100°C and 0°C
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Customers Who Bought This Also Bought