Porcelain Pestles
# de catalogue 470200-334
Fournisseur: SILICO & CHEMICO PORCELAIN SE
Spécifications
- Description:Porcelain Pestle, 114 mm, for 70 mm Mortar
- Cat. no.:470200-334
- Supplier No.:12091-259
Spécifications
A propos de cet article
Pestles for Porcelain Mortars.
The end of each pestle is unglazed.
Customers Who Bought This Also Bought