Low Form Porcelain Crucibles
# de catalogue 470200-314
Fournisseur: SILICO & CHEMICO PORCELAIN SE
Spécifications
- Description:Low Form Porcelain Crucibles, Size 2, 50 mL
- Cat. no.:470200-314
- Supplier No.:J236
Spécifications
A propos de cet article
These low, wide crucibles are made from highest quality porcelain. All surfaces are glazed except the outside bottom and rim. Without covers.
Customers Who Bought This Also Bought