Ward's® Live Paramecium multimicronucleatum Culture
# de catalogue 470199-682
Fournisseur: Avantor
Spécifications
- Description:P. multimicronucleatum (Small)
- Storage Conditions:Room temp, loose cap
- No. of Students:30
- Cat. no.:470199-682
- Packaging:Tube
- Media:Hay culture medium
- Supplier No.:WARD470199-682
Spécifications
A propos de cet article
Paramecium multimicronucleatumis one of the largest species of Paramecium. This ciliate contains a single macronucleus and four or more micronuclei. Grows in Dilute Alfalfa medium (Hay Medium).
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought